Assay Details
Gene Symbol: GEM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213579.1 | NP_998744.1 | 3 - 4 | 491 | 66 | |
GenBank mRNA | AB105812.1 | - | 3 - 4 | 491 | 66 | |
KY747399.1 | - | 2 - 3 | 434 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
