Assay Details
Gene Symbol: MUSTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213580.2 | NP_998745.1 | 2 - 3 | 206 | 119 | |
GenBank mRNA | AB040149.1 | - | 2 - 3 | 207 | 119 | |
BX930280.1 | - | 2 - 3 | 362 | 119 | ||
BX950259.2 | - | 2 - 3 | 366 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
