Assay Details
Gene Symbol: TBX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030367.1 | NP_001025538.1 | 4 - 5 | 571 | 65 | |
XM_025155425.1 | XP_025011193.1 | 6 - 7 | 894 | 65 | ||
GenBank mRNA | AB193180.1 | - | 4 - 5 | 571 | 65 | |
U67088.1 | - | 4 - 5 | 565 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
