Assay Details
Gene Symbol: TRH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030383.2 | NP_001025554.1 | 2 - 3 | 339 | 144 | |
XM_015293042.2 | XP_015148528.1 | 5 - 6 | 2137 | 144 | ||
XM_025154454.1 | XP_025010222.1 | 4 - 5 | 1264 | 144 | ||
GenBank mRNA | AB205116.1 | - | 2 - 3 | 339 | 144 | |
AJ703806.2 | - | 2 - 3 | 339 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
