Assay Details
Gene Symbol: KIAA1191 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030610.1 | NP_001025781.1 | 1 - 2 | 399 | 65 | |
XM_015293761.2 | XP_015149247.1 | 2 - 3 | 352 | 65 | ||
GenBank mRNA | AJ851782.1 | - | 1 - 2 | 399 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
