Assay Details
Gene Symbol: THG1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030616.1 | NP_001025787.1 | 3 - 4 | 523 | 98 | |
XM_015293767.1 | XP_015149253.1 | 3 - 4 | 474 | 98 | ||
XM_015293768.1 | XP_015149254.1 | 2 - 3 | 139 | 98 | ||
GenBank mRNA | AJ720720.1 | - | 3 - 4 | 523 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
