Assay Details
| Gene Symbol: SAR1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030621.1 | NP_001025792.1 | 6 - 7 | 580 | 79 | |
| XM_015294016.2 | XP_015149502.1 | 6 - 7 | 738 | 79 | ||
| XM_025154854.1 | XP_025010622.1 | 6 - 7 | 524 | 79 | ||
| XM_025154855.1 | XP_025010623.1 | 6 - 7 | 531 | 79 | ||
| GenBank mRNA | AJ720124.1 | - | 6 - 7 | 580 | 79 | |
| CR390339.1 | - | 6 - 7 | 532 | 79 | ||
More Information
Set Membership: |
|

SAR1B
Genomic Map