Assay Details
| Gene Symbol: HSPA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030793.3 | NP_001025964.2 | 3 - 4 | 626 | 65 | |
| GenBank mRNA | AJ851403.1 | - | 3 - 4 | 626 | 65 | |
| BM491256.1 | - | 3 - 4 | 596 | 65 | ||
More Information
Set Membership: |
|

HSPA13
Genomic Map