Assay Details
| Gene Symbol: EIF1AX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030804.1 | NP_001025975.1 | 2 - 3 | 266 | 103 | |
| GenBank mRNA | AJ720842.1 | - | 2 - 3 | 266 | 103 | |
More Information
Set Membership: |
|

EIF1AX
Genomic Map