Assay Details
| Gene Symbol: PIGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030808.1 | NP_001025979.1 | 4 - 5 | 1129 | 86 | |
| GenBank mRNA | AJ851372.1 | - | 4 - 5 | 1129 | 86 | |
More Information
Set Membership: |
|

PIGA
Genomic Map