Assay Details
Gene Symbol: STK24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030817.1 | NP_001025988.1 | 8 - 9 | 1162 | 69 | |
XM_015274308.1 | XP_015129794.1 | 7 - 8 | 1315 | 69 | ||
XM_025150144.1 | XP_025005912.1 | 9 - 10 | 1572 | 69 | ||
XM_025150145.1 | XP_025005913.1 | 9 - 10 | 1188 | 69 | ||
XM_025150149.1 | XP_025005917.1 | 10 - 11 | 1329 | 69 | ||
XM_025150154.1 | XP_025005922.1 | 8 - 9 | 986 | 69 | ||
XM_025150155.1 | XP_025005923.1 | 6 - 7 | 1068 | 69 | ||
GenBank mRNA | AJ851775.1 | - | 8 - 9 | 1162 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
