Assay Details
Gene Symbol: SUGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030823.2 | NP_001025994.2 | 12 - 13 | 964 | 68 | |
XM_015275411.2 | XP_015130897.1 | 12 - 13 | 909 | 68 | ||
GenBank mRNA | AJ720706.1 | - | 11 - 12 | 1039 | 68 | |
BU471834.1 | - | 2 - 3 | 290 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
