Assay Details
| Gene Symbol: RBBP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030914.2 | NP_001026085.1 | 393 | 87 | ||
| XM_015298921.2 | XP_015154407.1 | 393 | 87 | |||
| XM_015298922.2 | XP_015154408.1 | 393 | 87 | |||
| XM_015298923.2 | XP_015154409.1 | 393 | 87 | |||
| XM_025143689.1 | XP_024999457.1 | 333 | 87 | |||
| GenBank mRNA | AJ719611.1 | - | 395 | 87 | ||
More Information
Set Membership: |
|

RBBP5
Genomic Map