Assay Details
| Gene Symbol: BAK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030920.1 | NP_001026091.1 | 553 | 71 | ||
| GenBank mRNA | AJ851496.1 | - | 553 | 71 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BAK1| Gene Symbol: BAK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030920.1 | NP_001026091.1 | 553 | 71 | ||
| GenBank mRNA | AJ851496.1 | - | 553 | 71 | ||
Set Membership: |
|