Assay Details
Gene Symbol: DDX49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030938.1 | NP_001026109.1 | 10 - 11 | 1144 | 91 | |
XM_015300051.2 | XP_015155537.1 | 10 - 11 | 1135 | 91 | ||
GenBank mRNA | AJ719826.1 | - | 10 - 11 | 1144 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
