Assay Details
Gene Symbol: USE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030939.1 | NP_001026110.1 | 2 - 3 | 168 | 64 | |
XM_015300057.2 | XP_015155543.1 | 2 - 3 | 234 | 64 | ||
GenBank mRNA | AJ719267.1 | - | 2 - 3 | 168 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
