Assay Details
Gene Symbol: MC5R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031015.1 | NP_001026186.1 | 1 | 653 | 59 | |
XM_015282465.2 | XP_015137951.1 | 2 | 5145 | 59 | ||
XM_015282466.1 | XP_015137952.1 | 2 | 901 | 59 | ||
GenBank mRNA | KF670718.1 | - | 1 | 653 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
