Assay Details
| Gene Symbol: TIMM17B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031026.1 | NP_001026197.1 | 219 | 90 | ||
| XM_015298703.2 | XP_015154189.1 | 235 | 90 | |||
| XM_025143709.1 | XP_024999477.1 | 235 | 90 | |||
| GenBank mRNA | AJ720479.1 | - | 219 | 90 | ||
More Information
Set Membership: |
|

TIMM17B
Genomic Map