Assay Details
Gene Symbol: ACBD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031043.1 | NP_001026214.1 | 4 - 5 | 745 | 61 | |
XM_025148668.1 | XP_025004436.1 | 5 - 6 | 5264 | 61 | ||
GenBank mRNA | AJ720620.1 | - | 4 - 5 | 745 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
