Assay Details
| Gene Symbol: DUSP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031044.1 | NP_001026215.1 | 3 - 4 | 1315 | 85 | |
| GenBank mRNA | AJ720476.1 | - | 3 - 4 | 1315 | 85 | |
More Information
Set Membership: |
|

DUSP10
Genomic Map