Assay Details
| Gene Symbol: YIPF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031058.1 | NP_001026229.1 | 2 - 3 | 404 | 99 | |
| GenBank mRNA | AJ720497.1 | - | 2 - 3 | 404 | 99 | |
More Information
Set Membership: |
|

YIPF4
Genomic Map