Assay Details
| Gene Symbol: POMC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031098.1 | NP_001026269.1 | 1 - 2 | 130 | 134 | |
| XM_015285103.2 | XP_015140589.1 | 2 - 3 | 227 | 134 | ||
| GenBank mRNA | AB593424.1 | - | 2 - 3 | 384 | 134 | |
| AB593425.1 | - | 3 - 4 | 531 | 134 | ||
| AB593426.1 | - | 2 - 3 | 225 | 134 | ||
| AB593427.1 | - | 3 - 4 | 372 | 134 | ||
More Information
Set Membership: |
|

POMC
Genomic Map