Assay Details
Gene Symbol: USP12-like | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307997.3 | NP_001294926.1 | 2 - 3 | 249 | 75 | |
GenBank mRNA | AJ851557.1 | - | 2 - 3 | 208 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
