Assay Details
Gene Symbol: USP12-like | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ851557.1 | - | 5 - 6 | 738 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
