Assay Details
Gene Symbol: USP12-like | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307997.3 | NP_001294926.1 | 5 - 6 | 763 | 111 | |
XM_015278291.2 | XP_015133777.1 | 5 - 6 | 4760 | 111 | ||
XM_015278292.2 | XP_015133778.1 | 3 - 4 | 384 | 111 | ||
GenBank mRNA | AJ851557.1 | - | 6 - 7 | 808 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
