Assay Details
| Gene Symbol: NPY2R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031128.1 | NP_001026299.1 | 1 | 1052 | 99 | |
| XM_015285284.2 | XP_015140770.1 | 2 | 1122 | 99 | ||
| XM_015285285.2 | XP_015140771.1 | 3 | 1254 | 99 | ||
More Information
Set Membership: |
|

NPY2R
Genomic Map