Assay Details
| Gene Symbol: POMK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031132.1 | NP_001026303.1 | 1 - 2 | 467 | 94 | |
| GenBank mRNA | AJ851801.1 | - | 1 - 2 | 467 | 94 | |
More Information
Set Membership: |
|

POMK
Genomic Map