Assay Details
Gene Symbol: TMEM129 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031168.1 | NP_001026339.1 | 2 - 3 | 747 | 64 | |
GenBank mRNA | AJ720959.1 | - | 2 - 3 | 747 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
