Assay Details
| Gene Symbol: CACUL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031238.1 | NP_001026409.1 | 8 - 9 | 1147 | 66 | |
| GenBank mRNA | AJ719355.1 | - | 8 - 9 | 1147 | 66 | |
| CR353948.1 | - | 6 - 7 | 717 | 66 | ||
More Information
Set Membership: |
|

CACUL1
Genomic Map