Assay Details
| Gene Symbol: NEMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031242.1 | NP_001026413.1 | 3 - 4 | 458 | 102 | |
| GenBank mRNA | AJ720295.1 | - | 3 - 4 | 458 | 102 | |
More Information
Set Membership: |
|

NEMP2
Genomic Map