Assay Details
Gene Symbol: TRIM14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031431.1 | NP_001026602.1 | 2 - 3 | 298 | 89 | |
GenBank mRNA | AJ720736.1 | - | 2 - 3 | 298 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
