Assay Details
Gene Symbol: PUM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031437.1 | NP_001026608.1 | 13 - 14 | 1357 | 61 | |
XM_015280098.2 | XP_015135584.1 | 14 - 15 | 1519 | 61 | ||
XM_015280099.2 | XP_015135585.1 | 14 - 15 | 1485 | 61 | ||
XM_015280100.2 | XP_015135586.1 | 14 - 15 | 1459 | 61 | ||
XM_015280101.2 | XP_015135587.1 | 14 - 15 | 1453 | 61 | ||
GenBank mRNA | AJ719494.1 | - | 13 - 14 | 1394 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
