Assay Details
| Gene Symbol: CD72L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031446.2 | NP_001026617.2 | 6 - 7 | 909 | 66 | |
| GenBank mRNA | AB159677.1 | - | 6 - 7 | 828 | 66 | |
| AJ721072.1 | - | 6 - 7 | 913 | 66 | ||
| CR353309.1 | - | 4 - 5 | 620 | 66 | ||
More Information
Set Membership: |
|

CD72L1
Genomic Map