Assay Details
Gene Symbol: WDR70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031450.1 | NP_001026621.1 | 3 - 4 | 198 | 65 | |
NM_001195002.1 | NP_001181931.1 | 3 - 4 | 196 | 65 | ||
XM_015277332.2 | XP_015132818.1 | 3 - 4 | 245 | 65 | ||
GenBank mRNA | AJ720658.1 | - | 3 - 4 | 198 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
