Assay Details
Gene Symbol: PMEPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031492.1 | NP_001026663.1 | 2 - 3 | 427 | 66 | |
XM_025142264.1 | XP_024998032.1 | 3 - 4 | 380 | 66 | ||
XM_025142265.1 | XP_024998033.1 | 3 - 4 | 380 | 66 | ||
GenBank mRNA | AJ720618.1 | - | 2 - 3 | 427 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
