Assay Details
Gene Symbol: SSTR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031518.1 | NP_001026689.1 | 1 | 847 | 62 | |
XM_015283378.2 | XP_015138864.1 | 2 | 1429 | 62 | ||
GenBank mRNA | DQ069274.1 | - | 1 | 847 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
