Assay Details
Gene Symbol: NONO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031532.1 | NP_001026703.1 | 4 - 5 | 725 | 65 | |
XM_015278310.2 | XP_015133796.1 | 4 - 5 | 847 | 65 | ||
XM_015278311.2 | XP_015133797.1 | 4 - 5 | 819 | 65 | ||
XM_015278312.2 | XP_015133798.1 | 3 - 4 | 711 | 65 | ||
XM_015278313.2 | XP_015133799.1 | 4 - 5 | 731 | 65 | ||
XM_025149746.1 | XP_025005514.1 | 4 - 5 | 732 | 65 | ||
GenBank mRNA | AJ720639.1 | - | 4 - 5 | 725 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
