Assay Details
| Gene Symbol: MCCC2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031565.1 | NP_001026736.1 | 10 - 11 | 1650 | 76 | |
| XM_025152899.1 | XP_025008667.1 | 10 - 11 | 1653 | 76 | ||
| GenBank mRNA | AJ719309.1 | - | 10 - 11 | 1650 | 76 | |
More Information
Set Membership: |
|

MCCC2L
Genomic Map