Assay Details
| Gene Symbol: NDUFV1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031583.1 | NP_001026754.1 | 2 - 3 | 171 | 91 | |
| GenBank mRNA | AJ721062.1 | - | 2 - 3 | 171 | 91 | |
| CR354353.1 | - | 2 - 3 | 150 | 91 | ||
More Information
Set Membership: |
|

NDUFV1
Genomic Map