Assay Details
| Gene Symbol: FZD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030337.1 | NP_001025508.1 | 1 | 1751 | 120 | |
| GenBank mRNA | AF031830.1 | - | 1 | 1751 | 120 | |
| AF224314.1 | - | 1 | 622 | 120 | ||
More Information
Set Membership: |
|

FZD1
Genomic Map