Assay Details
| Gene Symbol: IL21R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030640.1 | NP_001025811.1 | 6 - 7 | 863 | 57 | |
| XM_025155100.1 | XP_025010868.1 | 6 - 7 | 853 | 57 | ||
| GenBank mRNA | AJ851394.1 | - | 6 - 7 | 863 | 57 | |
More Information
Set Membership: |
|

IL21R
Genomic Map