Assay Details
Gene Symbol: TMC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030641.2 | NP_001025812.2 | 3 - 4 | 495 | 72 | |
XM_015294224.2 | XP_015149710.1 | 3 - 4 | 653 | 72 | ||
XM_025155107.1 | XP_025010875.1 | 3 - 4 | 656 | 72 | ||
GenBank mRNA | AY581312.1 | - | 5 - 6 | 908 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
