Assay Details
Gene Symbol: UBE2H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030647.1 | NP_001025818.1 | 6 - 7 | 651 | 78 | |
GenBank mRNA | AJ719747.1 | - | 6 - 7 | 651 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
