Assay Details
| Gene Symbol: SNAP29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030652.1 | NP_001025823.1 | 1 - 2 | 261 | 84 | |
| XM_015275227.2 | XP_015130713.1 | 1 - 2 | 276 | 84 | ||
| GenBank mRNA | AJ719634.1 | - | 1 - 2 | 261 | 84 | |
More Information
Set Membership: |
|

SNAP29
Genomic Map