Assay Details
| Gene Symbol: TRIM41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030672.2 | NP_001025843.1 | 5 - 6 | 1124 | 65 | |
| XM_015294958.2 | XP_015150444.1 | 5 - 6 | 1479 | 65 | ||
More Information
Set Membership: |
|

TRIM41
Genomic Map