Assay Details
Gene Symbol: FAM18B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030699.1 | NP_001025870.1 | 2 - 3 | 125 | 103 | |
GenBank mRNA | AJ720440.1 | - | 2 - 3 | 125 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() |
