Assay Details
| Gene Symbol: FAM18B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030699.1 | NP_001025870.1 | 4 - 5 | 361 | 104 | |
| GenBank mRNA | AJ720440.1 | - | 4 - 5 | 361 | 104 | |
More Information
Set Membership: |
|

FAM18B1
Genomic Map