Assay Details
| Gene Symbol: COX10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030700.1 | NP_001025871.1 | 2 - 3 | 224 | 85 | |
| GenBank mRNA | AJ720296.1 | - | 2 - 3 | 224 | 85 | |
More Information
Set Membership: |
|

COX10
Genomic Map