Assay Details
Gene Symbol: SEPTIN9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030701.1 | NP_001025872.1 | 3 - 4 | 1007 | 94 | |
XM_015295143.2 | XP_015150629.1 | 3 - 4 | 1111 | 94 | ||
XM_015295144.2 | XP_015150630.1 | 4 - 5 | 1180 | 94 | ||
XM_015295145.2 | XP_015150631.1 | 3 - 4 | 1569 | 94 | ||
XM_015295146.2 | XP_015150632.1 | 3 - 4 | 1039 | 94 | ||
XM_015295147.2 | XP_015150633.1 | 2 - 3 | 238 | 94 | ||
XM_015295148.2 | XP_015150634.1 | 2 - 3 | 312 | 94 | ||
XM_015295149.2 | XP_015150635.1 | 2 - 3 | 337 | 94 | ||
XM_025141592.1 | XP_024997360.1 | 4 - 5 | 1301 | 94 | ||
GenBank mRNA | AJ851568.1 | - | 3 - 4 | 1007 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
