Assay Details
Gene Symbol: SEPTIN9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030701.1 | NP_001025872.1 | 9 - 10 | 1670 | 103 | |
XM_015295143.2 | XP_015150629.1 | 9 - 10 | 1774 | 103 | ||
XM_015295144.2 | XP_015150630.1 | 10 - 11 | 1843 | 103 | ||
XM_015295145.2 | XP_015150631.1 | 9 - 10 | 2232 | 103 | ||
XM_015295146.2 | XP_015150632.1 | 9 - 10 | 1702 | 103 | ||
XM_015295147.2 | XP_015150633.1 | 8 - 9 | 901 | 103 | ||
XM_015295148.2 | XP_015150634.1 | 8 - 9 | 975 | 103 | ||
XM_015295149.2 | XP_015150635.1 | 8 - 9 | 1000 | 103 | ||
XM_025141592.1 | XP_024997360.1 | 10 - 11 | 1964 | 103 | ||
GenBank mRNA | AJ851568.1 | - | 9 - 10 | 1670 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
